Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040106.2 | NP_001035195.1 | 7 | 1397 | ||
NM_177762.6 | NP_808430.2 | 7 | 1397 | |||
GenBank mRNA | AK147363.1 | - | 7 | 1398 | ||
AK158879.1 | - | 7 | 1287 | |||
BC141176.1 | - | 7 | 1153 | 9055788 |