Search
Search

Ogfod1| Gene Symbol: Ogfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093757.1 | NP_001087226.1 | 5 | 645 | ||
| NM_177767.4 | NP_808435.3 | 5 | 645 | |||
| GenBank mRNA | AK029629.1 | - | 5 | 647 | ||
| AK122532.1 | - | 5 | 549 | |||
| AK156765.1 | - | 5 | 615 | |||
| AK164189.1 | - | 5 | 532 | |||
| AK166343.1 | - | 5 | 568 | |||
| BB615031.1 | - | 5 | 646 | |||
| BC040267.1 | - | 5 | 605 | 5364674 | ||