Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173451.3 | NP_775627.1 | 2 | 2127 | ||
GenBank mRNA | AK034454.1 | - | 2 | 2128 | ||
AK034584.1 | - | 2 | 2151 | |||
AK046410.1 | - | 2 | 2111 | |||
AK052931.1 | - | 2 | 2108 | |||
BC117813.2 | - | 2 | 1347 | 40092671 |