Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTGS2 SIRNA Assays ›
          Gene Symbol:
          PTGS2
          Gene Name:
          prostaglandin-endoperoxide synthase 2
          Gene Aliases:
          COX-2, COX2, GRIPGHS, PGG/HS, PGHS-2, PHS-2, hCox-2
          Chromosome Location:
          Chr.1: 186671812 - 186680427 on Build GRCh38
          UniGene ID:
          Hs.196384
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11473

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTGS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000963.3 NP_000954.1 2 260
          GenBank mRNA AJ634912.1 - 2 123
          AK292167.1 - 2 248
          AY151286.1 - 2 220
          AY462100.1 - 2 123
          BC013734.1 - 2 243 3880850
          DA277667.1 - 2 260
          HM473188.1 - 1 40
          L15326.1 - 2 215
          M90100.1 - 2 220

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          angiogenesis
          prostaglandin metabolic process
          nicotinamide metabolic process
          movement of cell or subcellular component
          inflammatory response
          response to oxidative stress
          embryo implantation
          learning
          memory
          regulation of blood pressure
          negative regulation of cell proliferation
          response to fructose
          response to manganese ion
          response to lithium ion
          positive regulation of vascular endothelial growth factor production
          sensory perception of pain
          cyclooxygenase pathway
          lipoxygenase pathway
          bone mineralization
          ovulation
          positive regulation of prostaglandin biosynthetic process
          positive regulation of fever generation
          positive regulation of synaptic plasticity
          negative regulation of synaptic transmission, dopaminergic
          response to estradiol
          response to lipopolysaccharide
          response to vitamin D
          response to tumor necrosis factor
          cellular response to UV
          maintenance of blood-brain barrier
          positive regulation of NF-kappaB import into nucleus
          response to drug
          hair cycle
          positive regulation of apoptotic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of cell cycle
          positive regulation of vasoconstriction
          negative regulation of smooth muscle contraction
          positive regulation of smooth muscle contraction
          decidualization
          positive regulation of smooth muscle cell proliferation
          regulation of inflammatory response
          brown fat cell differentiation
          response to glucocorticoid
          negative regulation of calcium ion transport
          positive regulation of synaptic transmission, glutamatergic
          oxidation-reduction process
          response to fatty acid
          cellular response to mechanical stimulus
          cellular response to ATP
          cellular response to hypoxia
          cellular response to fluid shear stress
          positive regulation of transforming growth factor beta production
          positive regulation of cell migration involved in sprouting angiogenesis
          positive regulation of fibroblast growth factor production
          positive regulation of brown fat cell differentiation
          positive regulation of platelet-derived growth factor production
          cellular oxidant detoxification
          peroxidase activity
          prostaglandin-endoperoxide synthase activity
          protein binding
          lipid binding
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding
          arachidonate 15-lipoxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline