Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173037.1 | NP_766625.1 | 16 | 2903 | ||
GenBank mRNA | AK047206.1 | - | 11 | 1621 | ||
AK077757.1 | - | 16 | 2888 | |||
AK085030.1 | - | 16 | 2903 | |||
AK173250.1 | - | 16 | 2792 | |||
BC066134.1 | - | 16 | 2834 | |||
BC066869.1 | - | 16 | 2833 | 30106406 | ||
BC069838.1 | - | 16 | 2833 | 30106422 |