Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF5A SIRNA Assays ›
          Gene Symbol:
          Eif5a
          Gene Name:
          eukaryotic translation initiation factor 5A
          Gene Aliases:
          AA410058, D19Wsu54e, Eif4d, Eif5a1, eIF-4D, eIF-5A, eIF-5A-1, eIF-5A1
          Chromosome Location:
          Chr.11: 69916712 - 69921958 on Build GRCm38
          UniGene ID:
          Mm.18026
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s114770

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eif5a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166589.1 NP_001160061.1 4 568
          NM_001166590.1 NP_001160062.1 3 540
          NM_001166591.1 NP_001160063.1 4 504
          NM_001166592.1 NP_001160064.1 4 491
          NM_001166593.1 NP_001160065.1 4 417
          NM_001166594.1 NP_001160066.1 3 397
          NM_001166595.1 NP_001160067.1 3 339
          NM_001166596.1 NP_001160068.1 4 406
          NM_181582.4 NP_853613.1 3 474
          GenBank mRNA AK010183.1 - 3 375
          AK011306.1 - 3 430
          AK011470.1 - 3 359
          AK149705.1 - 3 428
          AK186286.1 - 2 124
          AK186900.1 - 2 128
          AK188240.1 - 2 154
          AK206538.1 - 2 152
          AY129323.1 - 4 417
          AY129324.1 - 3 540
          AY129325.1 - 3 474
          AY129326.1 - 4 568
          AY129327.1 - 4 504
          AY129328.1 - 3 390
          AY129329.1 - 4 484
          CX218195.1 - 3 397
          CX237346.1 - 4 480

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001099692, NM_001970, NM_001143761, NM_001143762, NM_001143760, and others.

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA export from nucleus
          translation
          translational elongation
          translational frameshifting
          protein export from nucleus
          transport
          apoptotic process
          positive regulation of cell proliferation
          protein transport
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of translational elongation
          positive regulation of translational termination
          mRNA transport
          positive regulation of muscle cell differentiation
          RNA binding
          translation elongation factor activity
          U6 snRNA binding
          ribosome binding
          poly(A) RNA binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-wb8cs:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline