Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF5A SIRNA Assays ›
          Gene Symbol:
          Eif5a
          Gene Name:
          eukaryotic translation initiation factor 5A
          Gene Aliases:
          AA410058, D19Wsu54e, Eif4d, Eif5a1, eIF-4D, eIF-5A, eIF-5A-1, eIF-5A1
          Chromosome Location:
          Chr.11: 69916712 - 69921958 on Build GRCm38
          UniGene ID:
          Mm.18026
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s114772

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eif5a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166589.1 NP_001160061.1 3 409
          NM_001166590.1 NP_001160062.1 2 381
          NM_001166591.1 NP_001160063.1 3 345
          NM_001166592.1 NP_001160064.1 3 332
          NM_001166593.1 NP_001160065.1 3 258
          NM_001166594.1 NP_001160066.1 2 238
          NM_001166595.1 NP_001160067.1 2 180
          NM_001166596.1 NP_001160068.1 3 247
          NM_181582.4 NP_853613.1 2 315
          GenBank mRNA AK010183.1 - 2 216
          AK011306.1 - 2 271
          AK011470.1 - 2 200
          AK149705.1 - 2 269
          AY129323.1 - 3 258
          AY129324.1 - 2 381
          AY129325.1 - 2 315
          AY129326.1 - 3 409
          AY129327.1 - 3 345
          AY129328.1 - 2 231
          AY129329.1 - 3 325
          BC003889.1 - 2 187 3591197
          BC008093.1 - 2 158 3587894
          BC024899.1 - 2 187 4482804
          CX218195.1 - 2 238
          CX237346.1 - 3 321

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001099692, NM_001970, NM_001143761, NM_001143762, NM_001143760, and others.
          This siRNA also targets the following transcripts in Rat: NM_001033681, XM_006246638, XM_006246637, XM_006246635, FQ220539, and others.

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA export from nucleus
          translation
          translational elongation
          translational frameshifting
          protein export from nucleus
          transport
          apoptotic process
          positive regulation of cell proliferation
          protein transport
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of translational elongation
          positive regulation of translational termination
          mRNA transport
          positive regulation of muscle cell differentiation
          RNA binding
          translation elongation factor activity
          U6 snRNA binding
          ribosome binding
          poly(A) RNA binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0