Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181545.4 | NP_853523.2 | 5 | 2134 | ||
GenBank mRNA | AK150875.1 | - | 3 | 1456 | ||
AY261798.1 | - | 5 | 2134 | |||
AY261799.1 | - | 5 | 2097 |
Gene Symbol: Slfn10-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041394.1 | - | 3 | 1268 | ||
AK144698.1 | - | 4 | 1861 | |||
AK220241.1 | - | 4 | 1985 | |||
AY261802.1 | - | 4 | 1860 | |||
AY261803.1 | - | 4 | 1829 | |||
BC115828.1 | - | 752 |
Gene Symbol: Slfn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172796.2 | NP_766384.2 | 4 | 2027 | ||
GenBank mRNA | AK036579.1 | - | 4 | 2028 | ||
AK050355.1 | - | 4 | 1981 | |||
AY217035.1 | - | 3 | 1495 | |||
AY261800.1 | - | 4 | 1935 | |||
AY261801.1 | - | 4 | 1917 |