Search
Search

Pced1a| Gene Symbol: Pced1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114541.1 | NP_001108013.1 | 7 | 1140 | ||
| NM_178762.4 | NP_848877.2 | 6 | 1727 | |||
| GenBank mRNA | AK031469.1 | - | 3 | 2858 | ||
| AK032354.1 | - | 6 | 1719 | |||
| AK032762.1 | - | 6 | 1702 | |||
| AK044590.1 | - | 3 | 2424 | |||
| AK045248.1 | - | 5 | 1797 | |||
| AK085613.1 | - | 5 | 1787 | |||
| AK180923.1 | - | 1 | 143 | |||
| AK213870.1 | - | 1 | 143 | |||
| BC064802.1 | - | 7 | 1186 | 30009464 | ||