Search
Search

Zfp182| Gene Symbol: Zfp182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013387.2 | NP_001013405.2 | 5 | 1259 | ||
| NM_001111076.1 | NP_001104546.1 | 5 | 1283 | |||
| GenBank mRNA | AK155039.1 | - | 5 | 1231 | ||
| BB574195.2 | - | 1 | 550 | |||
| BC063066.1 | - | 5 | 1224 | 6822685 | ||