Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CRISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131.2 | NP_001122.2 | 5 | 479 | ||
NM_001205220.1 | NP_001192149.1 | 5 | 490 | |||
NM_170609.1 | NP_733758.1 | 5 | 479 | |||
XM_011514315.2 | XP_011512617.1 | 2 | 610 | |||
XM_017010320.1 | XP_016865809.1 | 2 | 428 | |||
XM_017010321.1 | XP_016865810.1 | 2 | 138 | |||
GenBank mRNA | AB451334.1 | - | 4 | 400 | ||
AB451479.1 | - | 4 | 400 | |||
AK292497.1 | - | 5 | 460 | |||
BC131707.1 | - | 5 | 450 | |||
D38451.1 | - | 6 | 479 | |||
GQ472203.1 | - | 5 | 473 | |||
S80310.1 | - | 6 | 479 | |||
X95237.1 | - | 5 | 473 | |||
X95238.1 | - | 5 | 473 |