Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178767.5 | NP_848882.2 | 1 | 514 | ||
GenBank mRNA | AK039191.1 | - | 1 | 85 | ||
AK040702.1 | - | 1 | 515 | |||
AK050246.1 | - | 1 | 375 | |||
AK163155.1 | - | 1 | 295 | |||
BC038044.1 | - | 1 | 414 |