Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177059.3 | NP_796033.2 | 9 | 1325 | ||
GenBank mRNA | AF374459.1 | - | 9 | 1325 | ||
AK046350.1 | - | 9 | 1376 | |||
AK081114.1 | - | 9 | 1375 | |||
AK220367.1 | - | 5 | 705 | |||
BC144824.1 | - | 9 | 1295 |