Search
Search

PTPRJ| Gene Symbol: PTPRJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002843.3 | NP_002834.3 | 21 | 3700 | ||
| XM_017018083.1 | XP_016873572.1 | 20 | 3431 | |||
| XM_017018084.1 | XP_016873573.1 | 21 | 3813 | |||
| XM_017018085.1 | XP_016873574.1 | 21 | 3312 | |||
| GenBank mRNA | D37781.1 | - | 21 | 3689 | ||
| U10886.1 | - | 21 | 3694 | |||