Search
Search

Vstm4| Gene Symbol: Vstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178791.4 | NP_848906.1 | 8 | 1010 | ||
| GenBank mRNA | AK033244.1 | - | 8 | 966 | ||
| AK040699.1 | - | 8 | 858 | |||
| AK053685.1 | - | 8 | 1011 | |||
| BC107254.2 | - | 8 | 557 | |||
| BC125012.1 | - | 8 | 556 | |||
| BC153828.1 | - | 8 | 557 | |||