Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039105.3 | NP_001034194.1 | 1 | 98 | ||
NM_206974.1 | NP_996857.1 | 1 | 98 | |||
GenBank mRNA | AK053138.1 | - | 1 | 98 | ||
AY491736.1 | - | 1 | 73 | |||
AY491737.1 | - | 1 | 73 | |||
BC147091.1 | - | 1 | 66 | 9056140 | ||
BC147092.1 | - | 1 | 66 | 9056129 | ||
BC171997.1 | - | 1 | 66 | 9054283 |