Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045595.1 | - | 1 | 1393 | ||
BC110635.2 | - | 1 | 1132 | |||
BC125016.1 | - | 1 | 1134 |