Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PEX5 SIRNA Assays ›
Gene Symbol:
PEX5
Gene Name:
peroxisomal biogenesis factor 5
Gene Aliases:
PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
Chromosome Location:
Chr.12: 7188072 - 7218574 on Build GRCh38
UniGene ID:
Hs.567327
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11630
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PEX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000319.4
NP_000310.2
3
297
NM_001131023.1
NP_001124495.1
4
407
NM_001131024.1
NP_001124496.1
4
362
NM_001131025.1
NP_001124497.1
4
828
NM_001131026.1
NP_001124498.1
5
861
NM_001300789.1
NP_001287718.1
4
433
XM_005253455.1
XP_005253512.1
4
428
XM_011520793.2
XP_011519095.1
4
422
XM_011520795.1
XP_011519097.1
4
839
XM_011520797.1
XP_011519099.1
4
332
XM_011520798.1
XP_011519100.1
5
541
XM_011520799.2
XP_011519101.1
4
981
XM_011520800.1
XP_011519102.1
4
427
XM_011520801.2
XP_011519103.1
4
421
XM_011520802.2
XP_011519104.1
3
184
XM_017019744.1
XP_016875233.1
3
766
XM_017019745.1
XP_016875234.1
3
767
XM_017019746.1
XP_016875235.1
3
766
XM_017019747.1
XP_016875236.1
3
769
XM_017019748.1
XP_016875237.1
4
1030
XM_017019749.1
XP_016875238.1
4
794
XM_017019750.1
XP_016875239.1
4
332
XM_017019751.1
XP_016875240.1
4
443
XM_017019752.1
XP_016875241.1
5
541
XM_017019753.1
XP_016875242.1
4
427
XM_017019754.1
XP_016875243.1
4
638
XM_017019755.1
XP_016875244.1
4
1036
XM_017019756.1
XP_016875245.1
4
332
XM_017019757.1
XP_016875246.1
5
541
GenBank mRNA
AK225126.1
-
4
317
AK292256.1
-
5
861
AK299105.1
-
4
362
AK301700.1
-
1
78
AK302742.1
-
4
367
AK303515.1
-
4
433
AK312317.1
-
4
766
AK316250.1
-
4
317
BC010621.2
-
4
320
3872822
KU178292.1
-
3
281
KU178293.1
-
3
281
KU178294.1
-
3
281
KU178295.1
-
3
281
U19721.1
-
3
295
X84899.1
-
3
299
Z48054.1
-
4
367
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00165604_m1
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into peroxisome matrix
protein import into peroxisome matrix, docking
protein import into peroxisome membrane
negative regulation of protein homotetramerization
peroxisome targeting sequence binding
peroxisome matrix targeting signal-1 binding
protein binding
enzyme binding
small GTPase binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-jct7h:80/100.66.72.173:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0