Search
Search

Cpa6| Gene Symbol: Cpa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289497.1 | NP_001276426.1 | 4 | 597 | ||
| NM_177834.4 | NP_808502.2 | 5 | 722 | |||
| GenBank mRNA | AK078883.1 | - | 4 | 598 | ||
| AY773477.1 | - | 5 | 720 | |||
| BC119559.1 | - | 5 | 707 | 40057538 | ||
| BC119560.1 | - | 5 | 707 | 40057539 | ||