Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AGER SIRNA Assays ›
Gene Symbol:
AGER
Gene Name:
advanced glycosylation end product-specific receptor
Gene Aliases:
RAGE, SCARJ1
Chromosome Location:
Chr.6: 32180968 - 32185247 on Build GRCh38
UniGene ID:
Hs.534342
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1166
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AGER
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136.4
NP_001127.1
6
673
NM_001206929.1
NP_001193858.1
6
721
NM_001206932.1
NP_001193861.1
6
631
NM_001206934.1
NP_001193863.1
6
721
NM_001206936.1
NP_001193865.1
6
673
NM_001206940.1
NP_001193869.1
6
673
NM_001206954.1
NP_001193883.1
6
673
NM_001206966.1
NP_001193895.1
6
673
NM_172197.2
NP_751947.1
6
631
NR_038190.1
5
856
XM_017010328.1
XP_016865817.1
6
1646
GenBank mRNA
AB036432.1
-
6
597
AB061668.1
-
6
597
AB061669.1
-
5
780
AJ133822.1
-
6
546
AK298100.1
-
6
675
AK313178.1
-
6
601
AY755619.1
-
6
583
AY755620.1
-
6
583
AY755621.1
-
6
631
AY755622.1
-
6
631
AY755623.1
-
6
583
AY755624.1
-
6
541
AY755628.1
-
6
583
BC020669.1
-
6
592
4718076
DQ104251.1
-
6
583
DQ104252.1
-
6
541
DQ104253.1
-
6
583
DQ104254.1
-
5
766
EU117141.1
-
6
583
EU428812.1
-
5
756
EU428813.1
-
5
756
EU428815.1
-
5
756
EU428816.1
-
5
756
EU826618.1
-
6
573
EU826619.1
-
6
621
FJ545655.1
-
6
645
GU139370.1
-
3
1031
GU139371.1
-
3
1030
GU139372.1
-
5
830
GU139373.1
-
4
961
GU139374.1
-
4
907
GU139375.1
-
5
830
GU139376.1
-
5
766
GU139377.1
-
5
779
GU139378.1
-
6
595
HQ148133.1
-
6
597
HQ148134.1
-
6
561
HQ148135.1
-
5
777
KC692917.1
-
6
573
M91211.1
-
6
573
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
regulation of T cell mediated cytotoxicity
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
inflammatory response
negative regulation of cell adhesion
cell surface receptor signaling pathway
JAK-STAT cascade
brain development
glycoprotein metabolic process
response to wounding
response to fructose
positive regulation of autophagy
negative regulation of endothelial cell migration
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
positive regulation of fibroblast migration
response to activity
positive regulation of smooth muscle cell migration
lung development
neuron projection development
negative regulation of interleukin-10 production
positive regulation of interleukin-12 production
negative regulation of collagen biosynthetic process
response to vitamin A
response to genistein
negative regulation of osteoblast proliferation
cellular response to drug
positive regulation of activated T cell proliferation
positive regulation of neuron apoptotic process
innate immune response
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
positive regulation of inflammatory response
positive regulation of NF-kappaB transcription factor activity
response to methylglyoxal
calcium ion homeostasis
response to hyperoxia
positive regulation of phagocytosis, engulfment
transdifferentiation
cellular response to hydrogen peroxide
cellular response to glucose stimulus
cellular response to fatty acid
cellular response to organic cyclic compound
response to selenite ion
positive regulation of potassium ion transmembrane transporter activity
positive regulation of endothelial cell apoptotic process
positive regulation of reactive oxygen species metabolic process
regulation of CD4-positive, alpha-beta T cell activation
positive regulation of type B pancreatic cell apoptotic process
positive regulation of dendritic cell differentiation
receptor activity
transmembrane signaling receptor activity
protein binding
identical protein binding
S100 protein binding
advanced glycation end-product receptor activity
high mobility group box 1 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0