Search
Search

RAB2A| Gene Symbol: RAB2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242644.1 | NP_001229573.1 | 3 | 476 | ||
| NM_002865.2 | NP_002856.1 | 4 | 548 | |||
| GenBank mRNA | AF498930.1 | - | 4 | 250 | ||
| AK297576.1 | - | 3 | 450 | |||
| AK312344.1 | - | 4 | 471 | |||
| BC008929.2 | - | 4 | 438 | 2966694 | ||
| BT019695.1 | - | 4 | 250 | |||
| M28213.1 | - | 4 | 290 | |||
| X12953.1 | - | 4 | 458 | |||
| X13899.1 | - | 4 | 250 | |||