Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024673.2 | NP_001019844.2 | 1 | 120 | ||
GenBank mRNA | AY869695.1 | - | 1 | 120 | ||
BC150796.1 | - | 1 | 215 | 9087707 |
Gene Symbol: Ifnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177396.1 | NP_796370.1 | 1 | 120 | ||
GenBank mRNA | AY184375.1 | - | 1 | 120 | ||
AY869696.1 | - | 1 | 120 |