Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lama2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008481.2 | NP_032507.2 | 63 | 9097 | ||
GenBank mRNA | AK076479.1 | - | 8 | 1113 | ||
AK164427.1 | - | 1700 | ||||
AK220480.1 | - | 44 | 6303 | |||
BC025503.1 | - | 16 | 2172 | |||
BC039991.1 | - | 12 | 1570 | |||
BC054389.1 | - | 12 | 1576 | |||
U12147.1 | - | 63 | 9005 |