Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024917.1 | NP_001020088.1 | 11 | 3622 | ||
GenBank mRNA | AK136182.1 | - | 1 | 768 | ||
AK173167.1 | - | 9 | 3333 | |||
BC046458.1 | - | 2 | 1602 | |||
BC054549.1 | - | 11 | 3622 |