Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAD51 SIRNA Assays ›
          Gene Symbol:
          RAD51
          Gene Name:
          RAD51 recombinase
          Gene Aliases:
          BRCC5, FANCR, HRAD51, HsRad51, HsT16930, MRMV2, RAD51A, RECA
          Chromosome Location:
          Chr.15: 40694774 - 40732340 on Build GRCh38
          UniGene ID:
          Hs.631709
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11735

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAD51
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164269.1 NP_001157741.1 9 940
          NM_002875.4 NP_002866.2 9 1092
          NM_133487.3 NP_597994.3 9 1095
          XM_006720626.3 XP_006720689.1 9 1143
          XM_011521857.2 XP_011520159.1 9 842
          XM_011521858.2 XP_011520160.1 9 889
          XM_011521859.2 XP_011520161.1 9 1026
          XM_011521860.2 XP_011520162.1 9 937
          XM_011521862.2 XP_011520164.1 8 942
          GenBank mRNA AK131299.1 - 9 940
          AK291969.1 - 9 1050
          AK313503.1 - 9 1047
          BC001459.2 - 7 758 3139011
          BT019705.1 - 8 793
          CR536559.1 - 8 793
          D13804.1 - 9 1004
          D14134.1 - 9 1025

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          double-strand break repair via homologous recombination
          DNA recombinase assembly
          DNA synthesis involved in DNA repair
          strand displacement
          regulation of protein phosphorylation
          DNA unwinding involved in DNA replication
          DNA repair
          DNA recombination
          mitotic recombination
          cellular response to DNA damage stimulus
          meiotic nuclear division
          reciprocal meiotic recombination
          response to toxic substance
          response to X-ray
          regulation of double-strand break repair via homologous recombination
          telomere maintenance via telomere lengthening
          replication fork processing
          strand invasion
          response to drug
          positive regulation of DNA ligation
          protein homooligomerization
          chromosome organization involved in meiotic cell cycle
          cellular response to ionizing radiation
          cellular response to gamma radiation
          cellular response to hydroxyurea
          cellular response to cisplatin
          cellular response to camptothecin
          response to etoposide
          replication-born double-strand break repair via sister chromatid exchange
          mitotic recombination-dependent replication fork processing
          recombinase activity
          four-way junction DNA binding
          chromatin binding
          double-stranded DNA binding
          single-stranded DNA binding
          endodeoxyribonuclease activity
          protein binding
          ATP binding
          protein C-terminus binding
          identical protein binding
          single-stranded DNA-dependent ATPase activity
          DNA polymerase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0