siRNA Details
Gene Symbol: Rsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013769.1 | NP_001013791.1 | 4 | 682 | ||
GenBank mRNA | AK087838.1 | - | 1 | 306 | ||
BC103788.1 | - | 4 | 657 | 6333828 |

Gene Symbol: Rsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013769.1 | NP_001013791.1 | 4 | 682 | ||
GenBank mRNA | AK087838.1 | - | 1 | 306 | ||
BC103788.1 | - | 4 | 657 | 6333828 |