Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAD51D SIRNA Assays ›
          Gene Symbol:
          RAD51D RAD51L3-RFFL
          Gene Name:
          RAD51 paralog D
          RAD51L3-RFFL readthrough
          Gene Aliases:
          BROVCA4, R51H3, RAD51L3, TRAD
          -
          Chromosome Location:
          Chr.17: 35099792 - 35119869 on Build GRCh38
          Chromosome Location:
          Chr.17: 35009112 - 35121522 on Build GRCh38
          UniGene ID:
          Hs.631757
          Hs.13680
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s11744

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAD51D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142571.1 NP_001136043.1 6 827
          NM_002878.3 NP_002869.3 6 767
          NM_133629.2 NP_598332.1 3 431
          NR_037711.1 5 648
          NR_037712.1 4 513
          GenBank mRNA AB013341.1 - 6 617
          AB016223.1 - 5 498
          AB016224.1 - 4 363
          AB016225.1 - 3 281
          AB018360.1 - 4 279
          AB018361.1 - 3 197
          AB018362.1 - 4 376
          AB018363.1 - 6 593
          AB020412.1 - 1 21
          AF034956.1 - 5 518
          AK296241.1 - 6 771
          AK301249.1 - 6 833
          AK302450.1 - 6 661
          AL117459.1 - 2 48
          BC002723.2 - 5 532 3628519
          BC014422.1 - 6 646 4639227
          BI916871.1 - 6 767
          BX647297.1 - 6 714
          Y15572.1 - 6 630
          Gene Symbol: RAD51L3-RFFL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037714.1 2 263
          GenBank mRNA AK097811.1 - 2 234

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic DNA recombinase assembly
          telomere maintenance
          double-strand break repair via homologous recombination
          DNA synthesis involved in DNA repair
          strand displacement
          DNA repair
          nucleotide-excision repair
          mitotic recombination
          reciprocal meiotic recombination
          response to ionizing radiation
          interstrand cross-link repair
          strand invasion
          regulation of cell cycle
          recombinase activity
          four-way junction DNA binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          endodeoxyribonuclease activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          gamma-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0