Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287179.2 | NP_001274108.1 | 2, 3 | 1577 | ||
GenBank mRNA | AB001431.1 | - | 1, 2 | 36 | ||
AB731728.1 | - | 2, 3 | 1467 | |||
AK087451.1 | - | 3, 4 | 1606 | |||
BC158132.1 | - | 2, 3 | 1490 | 9007399 |