Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RANGAP1 SIRNA Assays ›
          Gene Symbol:
          RANGAP1
          Gene Name:
          Ran GTPase activating protein 1
          Gene Aliases:
          Fug1, RANGAP, SD
          Chromosome Location:
          Chr.22: 41244777 - 41302213 on Build GRCh38
          UniGene ID:
          Hs.183800
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11777

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RANGAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278651.1 NP_001265580.1 9, 10 1662
          NM_001317930.1 NP_001304859.1 8, 9 1116
          NM_002883.3 NP_002874.1 8, 9 1201
          XM_005261695.1 XP_005261752.1 8, 9 1329
          XM_005261696.1 XP_005261753.1 8, 9 1100
          XM_006724289.3 XP_006724352.1 8, 9 1104
          XM_011530293.1 XP_011528595.1 7, 8 1194
          XM_011530294.2 XP_011528596.1 8, 9 2810
          XM_011530295.2 XP_011528597.1 8, 9 1509
          XM_011530297.1 XP_011528599.1 8, 9 1329
          XM_017028893.1 XP_016884382.1 9, 10 2440
          XM_017028894.1 XP_016884383.1 9, 10 2578
          XM_017028895.1 XP_016884384.1 8, 9 2940
          XM_017028896.1 XP_016884385.1 9, 10 1328
          XM_017028897.1 XP_016884386.1 8, 9 2420
          XM_017028898.1 XP_016884387.1 10, 11 2353
          XM_017028899.1 XP_016884388.1 7, 8 1066
          GenBank mRNA AB058738.1 - 7, 8 2385
          AK302222.1 - 7, 8 765
          AK303524.1 - 8, 9 1124
          BC014044.2 - 8, 9 1116 3506039
          BC041396.2 - 9, 10 1672 5277434
          CR456557.1 - 7, 8 958
          X82260.1 - 7, 8 897

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          sister chromatid cohesion
          signal transduction
          protein sumoylation
          positive regulation of GTPase activity
          negative regulation of protein export from nucleus
          response to axon injury
          cell-cell adhesion
          cellular response to vasopressin
          GTPase activator activity
          protein binding
          Ran GTPase binding
          ubiquitin protein ligase binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-6cddc:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline