Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013774.2 | NP_001013796.2 | 8 | 907 | ||
GenBank mRNA | AK136027.1 | - | 4 | 425 | ||
AY950703.1 | - | 8 | 908 | |||
FJ717332.1 | - | 6 | 674 | |||
FJ717333.1 | - | 7 | 737 |