Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAP1GAP SIRNA Assays ›
Gene Symbol:
RAP1GAP
Gene Name:
RAP1 GTPase activating protein
Gene Aliases:
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location:
Chr.1: 21596215 - 21669444 on Build GRCh38
UniGene ID:
Hs.148178
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11786
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAP1GAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145657.1
NP_001139129.1
8
600
NM_001145658.1
NP_001139130.1
7
460
NM_002885.2
NP_002876.2
7
493
XM_005245955.3
XP_005246012.2
7
505
XM_006710804.2
XP_006710867.2
7
505
XM_006710805.2
XP_006710868.2
7
505
XM_017001965.1
XP_016857454.1
6
507
XM_017001966.1
XP_016857455.1
6
507
XM_017001967.1
XP_016857456.1
7
505
XM_017001968.1
XP_016857457.1
7
540
XM_017001969.1
XP_016857458.1
6
507
XM_017001970.1
XP_016857459.1
6
507
XM_017001971.1
XP_016857460.1
7
459
XM_017001972.1
XP_016857461.1
7
505
XM_017001973.1
XP_016857462.1
6
538
XM_017001974.1
XP_016857463.1
7
505
XM_017001975.1
XP_016857464.1
6
507
XM_017001976.1
XP_016857465.1
7
505
XM_017001977.1
XP_016857466.1
7
505
XM_017001978.1
XP_016857467.1
8
654
XM_017001979.1
XP_016857468.1
9
658
XM_017001980.1
XP_016857469.1
7
459
XM_017001981.1
XP_016857470.1
9
634
XM_017001982.1
XP_016857471.1
8
654
XM_017001983.1
XP_016857472.1
10
866
XM_017001984.1
XP_016857473.1
9
634
XM_017001985.1
XP_016857474.1
7
459
XM_017001986.1
XP_016857475.1
9
671
XM_017001987.1
XP_016857476.1
8
600
XM_017001988.1
XP_016857477.1
8
571
XM_017001989.1
XP_016857478.1
9
671
XM_017001990.1
XP_016857479.1
8
556
XM_017001991.1
XP_016857480.1
8
594
GenBank mRNA
AB003930.1
-
5
434
AB007943.3
-
8
538
BC035030.1
-
4
249
5261027
BC054490.1
-
7
400
5767775
BP200029.1
-
7
460
BP283134.1
-
7
493
CV574545.1
-
7
450
M64788.1
-
7
436
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
regulation of GTPase activity
positive regulation of GTPase activity
negative regulation of neuron differentiation
regulation of small GTPase mediated signal transduction
negative regulation of microvillus assembly
negative regulation of GTP binding
negative regulation of thyroid gland epithelial cell proliferation
cellular response to glial cell derived neurotrophic factor
GTPase activity
GTPase activator activity
protein binding
Ras GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0