Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IGK-V1 SIRNA Assays ›
          Gene Symbol:
          Igk-V1 Igh-V7183 Igk-V21 Igkc Igkv1-110 Igkv1-117 Igkv9-120 Zak
          Gene Name:
          immunoglobulin kappa chain variable 1 (V1)
          immunoglobulin heavy chain (V7183 family)
          immunoglobulin kappa chain variable 21 (V21)
          immunoglobulin kappa constant
          immunoglobulin kappa variable 1-110
          immunoglobulin kappa variable 1-117
          immunoglobulin kappa chain variable 9-120
          sterile alpha motif and leucine zipper containing kinase AZK
          Gene Aliases:
          -
          B9-scFv, IgG, IgH, IgVH1(VSG), VH7183, VI24H
          -
          Igk-C
          Gm16634, Igk, K5.1
          Cr1, Gm16797, IgK, Igk-V21, V(k)45.1
          EG434025, Gm5571
          AV006891, B230120H23Rik, MLTK, MLTKalpha, MLTKbeta
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.6: 70726435 - 70726754 on Build GRCm38
          Chromosome Location:
          Chr.6: 68270527 - 68271265 on Build GRCm38
          Chromosome Location:
          Chr.6: 68121091 - 68121825 on Build GRCm38
          Chromosome Location:
          Chr.6: 68049983 - 68050454 on Build GRCm38
          Chromosome Location:
          Chr.2: 72285637 - 72442610 on Build GRCm38
          UniGene ID:
          Mm.304143
          Mm.342177
          Mm.304143
          Mm.333124
          Mm.333124
          Mm.333124
          Mm.304143
          Mm.304143
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s117865

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Igk-V1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AF154883.1 - 1 60
          AF154884.1 - 1 60
          AF154885.1 - 1 60
          AF154888.1 - 1 60
          AF154889.1 - 1 60
          AF154890.1 - 1 60
          AF154891.1 - 1 60
          AF154892.1 - 1 60
          AF154893.1 - 1 60
          AF154894.1 - 1 60
          AF154895.1 - 1 60
          AF154896.1 - 1 60
          AF154897.1 - 1 60
          AF154898.1 - 1 60
          Z95477.1 - 156
          Gene Symbol: Igh-V7183
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U62527.1 - 624
          Gene Symbol: Igk-V21
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U29768.1 - 1 146
          X67624.1 - 1 153
          Gene Symbol: Igkc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U29147.1 - 153
          Z27396.1 - 153
          Gene Symbol: Igkv1-110
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA L39088.1 - 2 210
          M20828.1 - 2 233
          M32381.1 - 2 210
          M32382.1 - 2 210
          M32384.1 - 2 210
          Gene Symbol: Igkv1-117
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA M55313.1 - 2 243
          S64186.1 - 2 249
          X51720.1 - 2 229
          X66624.1 - 2 234
          X66628.1 - 2 247
          X66631.1 - 2 249
          X66637.1 - 2 248
          X66640.1 - 2 245
          Gene Symbol: Igkv9-120
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA Z95478.1 - 171
          Gene Symbol: Zak
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U29267.1 - 153

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-6brs2:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline