Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003920.3 | NP_001003920.2 | 5 | 672 | ||
NM_001168572.1 | NP_001162044.1 | 5 | 672 | |||
GenBank mRNA | AY533671.1 | - | 5 | 514 | ||
BC086636.1 | - | 5 | 671 | 6402113 | ||
EU016556.1 | - | 5 | 520 | |||
EU016557.1 | - | 5 | 514 | |||
EU016558.1 | - | 5 | 514 | |||
EU586326.1 | - | 5 | 702 |