Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033466.3 | NP_001028638.2 | 3 | 880 | ||
GenBank mRNA | AK030936.1 | - | 3 | 728 | ||
AK132308.1 | - | 3 | 881 | |||
AK160748.1 | - | 3 | 710 | |||
AK173184.1 | - | 1 | 4455 | |||
BC094553.1 | - | 1 | 308 |