Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193273.1 | NP_001180202.1 | 13 | 1587 | ||
NM_005057.3 | NP_005048.2 | 13 | 1643 | |||
XM_011509833.2 | XP_011508135.1 | 13 | 1641 | |||
GenBank mRNA | AK296978.1 | - | 13 | 1541 | ||
AK297540.1 | - | 12 | 1880 | |||
BC053856.1 | - | 13 | 1614 | 5576406 | ||
BC075059.2 | - | 13 | 1541 | 30915584 | ||
BC075060.2 | - | 13 | 1541 | 30915368 | ||
X85134.1 | - | 13 | 1609 |