Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198719.1 | NP_001185648.1 | 2 | 422 | ||
NM_002893.3 | NP_002884.1 | 2 | 434 | |||
GenBank mRNA | AK091911.1 | - | 2 | 419 | ||
AK291567.1 | - | 2 | 178 | |||
BC114500.1 | - | 2 | 332 | 40083852 | ||
BC114501.1 | - | 2 | 332 | 40083851 | ||
CR541701.1 | - | 2 | 76 | |||
DA924163.1 | - | 2 | 422 | |||
DB460068.1 | - | 2 | 437 | |||
U35143.1 | - | 2 | 337 | |||
X72841.1 | - | 2 | 361 |