Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207537.1 | NP_997420.1 | 1 | 130 | ||
GenBank mRNA | BC107192.1 | - | 1 | 129 | ||
BC107193.1 | - | 1 | 129 | |||
BC114400.1 | - | 1 | 129 | |||
BC114428.1 | - | 1 | 129 |
Set Membership: |
G-protein coupled receptor |