Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013785.3 | NP_001013807.2 | 3 | 363 | ||
GenBank mRNA | BC087964.1 | - | 3 | 363 | 6767816 | |
BG086571.2 | - | 469 |