Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024706.2 | NP_001019877.2 | 2 | 223 | ||
GenBank mRNA | BC060995.1 | - | 2 | 223 | 6774503 |
Gene Symbol: 4930555G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175393.4 | NP_780602.2 | 2 | 236 | ||
GenBank mRNA | AK076920.1 | - | 2 | 237 |
Gene Symbol: D830030K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177135.4 | NP_796109.1 | 2 | 81 | ||
GenBank mRNA | AK082998.1 | - | 2 | 158 | ||
AK085928.1 | - | 2 | 82 |
Gene Symbol: Gm10406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164727.1 | NP_001158199.1 | 4 | 358 | ||
GenBank mRNA | BC172015.1 | - | 3 | 237 | 9054301 | |
EF651804.1 | - | 2 | 154 | |||
EF651806.1 | - | 2 | 154 | |||
EF651807.1 | - | 2 | 154 |
Gene Symbol: Gm10408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256501.1 | NP_001243430.1 | 2 | 223 |
Gene Symbol: Gm10409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BY739387.1 | - | 381 |
Gene Symbol: Gm10413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029288.3 | NP_083564.2 | 2 | 217 | ||
GenBank mRNA | AK005583.1 | - | 2 | 207 | ||
BC100412.1 | - | 2 | 217 | 30916569 |
Gene Symbol: Gm2237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB056480.1 | - | 3 | 211 | ||
CX218842.1 | - | 1 | 239 |
Gene Symbol: Gm2897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177715.2 | NP_001171186.1 | 4 | 393 | ||
GenBank mRNA | BC028273.1 | - | 4 | 403 |
Gene Symbol: Gm3002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC080297.1 | - | 2 | 3622 |
Gene Symbol: Gm3020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012594.1 | - | 4 | 397 | ||
BY711739.1 | - | 4 | 397 |
Gene Symbol: Gm3317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242941.2 | NP_001229870.2 | 2 | 154 |
Gene Symbol: Gm3373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040090.1 | - | 545 | 3987869 | ||
BC096548.1 | - | 2030 |
Gene Symbol: Gm3383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291093.1 | NP_001278022.1 | 3 | 250 | ||
NM_001291094.1 | NP_001278023.1 | 62 | ||||
GenBank mRNA | EF651835.1 | - | 3 | 250 |
Gene Symbol: Gm3488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256885.1 | NP_001243814.1 | 2 | 154 |
Gene Symbol: Gm3500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256886.1 | NP_001243815.1 | 4 | 383 | ||
GenBank mRNA | EF651805.1 | - | 154 | |||
EF651808.1 | - | 154 | ||||
EF651809.1 | - | 154 |
Gene Symbol: Gm3558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270842.1 | NP_001257771.1 | 4 | 457 | ||
GenBank mRNA | BC089473.1 | - | 4 | 457 | 6588560 |
Gene Symbol: Gm3636 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN668200.1 | - | 2 | 135 |
Gene Symbol: Gm3696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024712.2 | NP_001019883.2 | 358 | |||
GenBank mRNA | BC055874.1 | - | 357 | 3491523 |
Gene Symbol: Gm5795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270806.1 | NP_001257735.1 | 2 | 188 | ||
GenBank mRNA | BU946301.1 | - | 2 | 191 |
Gene Symbol: Gm5796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029930.2 | NP_001025101.2 | 216 | |||
GenBank mRNA | BC093494.1 | - | 216 | 2812218 |
Gene Symbol: Hn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC055351.1 | - | 480 |
Gene Symbol: LOC100036572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027416.1 | - | 1153 |
Gene Symbol: LOC100861615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270812.1 | NP_001257741.1 | 3 | 209 | ||
GenBank mRNA | AK049619.1 | - | 3 | 210 |
Gene Symbol: LOC545005 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089308.1 | - | 1 | 940 |