Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENO1B SIRNA Assays ›
          Gene Symbol:
          Eno1b Eno1
          Gene Name:
          enolase 1B, retrotransposed
          enolase 1, alpha non-neuron
          Gene Aliases:
          EG433182, Eno1, Gm5506
          0610008I15, AL022784, Eno-1, MBP-1
          Chromosome Location:
          Chr.18: 48045335 - 48048379 on Build GRCm38
          Chromosome Location:
          Chr.4: 150236726 - 150248873 on Build GRCm38
          UniGene ID:
          Mm.372357
          Mm.372357
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s119371

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eno1b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025388.1 NP_001020559.1 2 379
          GenBank mRNA BC089539.1 - 2 379 6707317
          Gene Symbol: Eno1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_023119.2 NP_075608.2 4, 5 397
          GenBank mRNA AK002322.1 - 4, 5 356
          AK002336.1 - 4, 5 355
          AK088788.1 - 4, 5 341
          AK147987.1 - 4, 5 355
          AK150161.1 - 4, 5 355
          AK150192.1 - 4, 5 356
          AK153321.1 - 4, 5 354
          AK180361.1 - 2, 3 79
          AK191162.1 - 1, 2 79
          AK191332.1 - 2, 3 79
          AK193395.1 - 79
          AK196981.1 - 2, 3 79
          AK198034.1 - 2, 3 79
          AK199229.1 - 2, 3 79
          AK202164.1 - 2, 3 79
          AK202428.1 - 2, 3 79
          AK202509.1 - 1, 2 57
          AK204087.1 - 2, 3 79
          AK204457.1 - 2, 3 79
          AK204949.1 - 2, 3 79
          AK214077.1 - 3, 4 213
          BC003891.1 - 4, 5 347 3592683
          BC003934.1 - 4, 5 369
          BC010685.1 - 4, 5 335 3599051
          BC024644.1 - 4, 5 327 3583123
          BC039179.1 - 4, 5 317 4501041
          BC083334.1 - 4, 5 329 6414729
          BC085098.1 - 3, 4 258 5376359
          BU526382.1 - 4, 5 400
          X52379.1 - 4, 5 339

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lyase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycolytic process
          in utero embryonic development
          positive regulation of binding
          cellular response to acid chemical
          phosphopyruvate hydratase activity
          protein binding
          protein homodimerization activity
          poly(A) RNA binding
          protein heterodimerization activity
          GTPase binding
          RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0