Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHOA SIRNA Assays ›
          Gene Symbol:
          Rhoa
          Gene Name:
          ras homolog gene family, member A
          Gene Aliases:
          Arha, Arha1, Arha2
          Chromosome Location:
          Chr.9: 108306205 - 108337939 on Build GRCm38
          UniGene ID:
          Mm.318359
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s119553

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rhoa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016802.4 NP_058082.2 6 931
          GenBank mRNA AF014371.1 - 5 639
          AF178958.1 - 5 526
          AF178959.1 - 5 526
          AF178960.1 - 5 526
          AF178961.1 - 5 526
          AK077606.1 - 6 739
          AK083624.1 - 6 738
          AK150294.1 - 6 775
          AK150475.1 - 6 782
          AK152629.1 - 6 718
          AK165512.1 - 6 714
          AK172051.1 - 6 739
          AK191457.1 - 1 93
          BC068115.1 - 6 931 6809149
          BC096423.1 - 6 717 3590185
          JN971019.1 - 4 505

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001664, L25080, AK307220, AF498970, AK130066, and others.

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          regulation of transcription from RNA polymerase II promoter
          cytoskeleton organization
          cell cycle
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          small GTPase mediated signal transduction
          Rho protein signal transduction
          skeletal muscle tissue development
          regulation of neuron projection development
          cerebral cortex cell migration
          forebrain radial glial cell differentiation
          actin cytoskeleton organization
          cell differentiation
          positive regulation of cell growth
          regulation of cell migration
          androgen receptor signaling pathway
          positive regulation of actin filament polymerization
          stress-activated protein kinase signaling cascade
          positive regulation of cytokinesis
          negative regulation of intracellular steroid hormone receptor signaling pathway
          regulation of osteoblast proliferation
          cleavage furrow formation
          apolipoprotein A-I-mediated signaling pathway
          response to drug
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          stress fiber assembly
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          apical junction assembly
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          ossification involved in bone maturation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of smooth muscle contraction
          GTP metabolic process
          regulation of dendrite development
          negative chemotaxis
          cell division
          positive regulation of stress fiber assembly
          regulation of calcium ion transport
          positive regulation of lipase activity
          negative regulation of cell death
          trabecula morphogenesis
          positive regulation of podosome assembly
          mitotic spindle assembly
          regulation of neural precursor cell proliferation
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          myosin binding
          GDP binding
          protein domain specific binding
          Rho GDP-dissociation inhibitor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qq54g:80/100.66.72.153:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0