Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081234.1 | NP_001074703.1 | 11 | 2326 | ||
GenBank mRNA | BC051059.1 | - | 5 | 1207 |