Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHD SIRNA Assays ›
Gene Symbol:
RHD
Gene Name:
Rh blood group D antigen
Gene Aliases:
CD240D, DIIIc, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPI
Chromosome Location:
Chr.1: 25272393 - 25330445 on Build GRCh38
UniGene ID:
Hs.449968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12016
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127691.2
NP_001121163.1
4
664
NM_001282867.1
NP_001269796.1
4
660
NM_001282868.1
NP_001269797.1
4
664
NM_001282869.1
NP_001269798.1
4
664
NM_001282870.1
NP_001269799.1
4
664
NM_001282871.1
NP_001269800.1
4
664
NM_001282872.1
NP_001269801.1
4
664
NM_016124.4
NP_057208.2
4
664
XM_017002015.1
XP_016857504.1
4
757
GenBank mRNA
AB018966.1
-
4
602
AB018967.1
-
4
602
AB018968.1
-
4
602
AB018969.1
-
4
602
AB037270.1
-
4
602
AB046420.1
-
4
602
AB049754.1
-
4
602
AF037626.1
-
4
598
AF177938.1
-
4
602
AF510069.1
-
4
645
AF510070.1
-
4
645
AJ704215.2
-
4
602
AK297151.1
-
4
757
AM183924.3
-
4
602
AY449381.1
-
4
602
AY449382.1
-
4
602
AY449383.1
-
4
602
AY449384.1
-
4
602
AY449385.1
-
4
602
AY751491.1
-
4
602
AY751492.1
-
4
602
AY751493.1
-
4
602
AY751494.1
-
4
602
AY751495.1
-
4
602
AY751496.1
-
4
602
BC139922.1
-
4
652
40148684
DQ451878.1
-
4
621
EU557240.1
-
4
605
EU557241.1
-
4
605
EU661905.1
-
4
542
EU661906.1
-
4
542
EU661907.1
-
1
95
FR745439.1
-
4
602
FR745440.1
-
4
602
FR745441.1
-
4
602
HE657775.1
-
4
602
JN635688.1
-
4
602
JQ065733.1
-
4
602
JQ424879.1
-
4
660
KT240043.1
-
4
602
L08429.1
-
4
602
LC053447.1
-
4
602
LN714468.1
-
4
602
LN824132.1
-
4
602
S57971.1
-
4
602
S70174.1
-
4
602
S73913.1
-
4
602
S78509.1
-
4
602
S82449.1
-
4
602
X63094.1
-
4
660
X63097.1
-
4
618
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
organic cation transport
nitrogen utilization
ammonium transmembrane transport
ammonium transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0