Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010888.2 | NP_035018.1 | 4 | 483 | ||
GenBank mRNA | AK147147.1 | - | 4 | 513 | ||
BC059730.1 | - | 4 | 456 | |||
BC076501.1 | - | 4 | 514 | |||
BC086933.1 | - | 4 | 456 | 6760537 | ||
BG081040.2 | - | 4 | 473 |