Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026533.3 | NP_080809.1 | 5 | 466 | ||
GenBank mRNA | AK012479.1 | - | 5 | 411 | ||
AK018703.1 | - | 5 | 410 | |||
AK076060.1 | - | 5 | 410 | |||
AK088237.1 | - | 4 | 1043 | |||
AK151043.1 | - | 5 | 411 | |||
AK151671.1 | - | 5 | 412 | |||
AK161798.1 | - | 3 | 1458 | |||
AK166691.1 | - | 5 | 411 | |||
AV498110.1 | - | 4 | 373 | |||
BC011192.1 | - | 5 | 409 | 4016679 | ||
BC085269.1 | - | 2 | 1564 | |||
BC090397.1 | - | 5 | 393 | 5150054 | ||
BC106137.1 | - | 5 | 394 | 4990874 | ||
BC125602.1 | - | 5 | 410 | 40130117 | ||
BC125604.1 | - | 5 | 410 | 40130119 |