Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS6KB1 SIRNA Assays ›
          Gene Symbol:
          RPS6KB1
          Gene Name:
          ribosomal protein S6 kinase B1
          Gene Aliases:
          PS6K, S6K, S6K-beta-1, S6K1, STK14A, p70 S6KA, p70(S6K)-alpha, p70-S6K, p70-alpha
          Chromosome Location:
          Chr.17: 59893046 - 59950426 on Build GRCh38
          UniGene ID:
          Hs.463642
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s12282
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPS6KB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001272042.1 NP_001258971.1 3 342
          NM_001272043.1 NP_001258972.1 3 342
          NM_001272044.1 NP_001258973.1 4 471
          NM_001272060.1 NP_001258989.1 3 342
          NM_003161.3 NP_003152.1 3 342
          XM_011525101.2 XP_011523403.1 3 337
          XM_011525102.2 XP_011523404.1 3 337
          XM_011525103.2 XP_011523405.1 4 391
          XM_017024929.1 XP_016880418.1 4 400
          XM_017024930.1 XP_016880419.1 3 301
          XM_017024931.1 XP_016880420.1 3 262
          XM_017024932.1 XP_016880421.1 3 262
          XM_017024933.1 XP_016880422.1 3 257
          GenBank mRNA AK293247.1 - 3 290
          AK297147.1 - 4 278
          AK300202.1 - 3 258
          AK312875.1 - 3 248
          BC036033.1 - 3 302
          BC053365.1 - 3 244 6142009
          BP359158.1 - 3 342
          M60724.1 - 3 230

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031985, XM_008768121, XM_008768120, XM_006247111, M58340, and others.
          This siRNA also targets the following transcripts in Mouse: NM_028259, NM_001114334, AK178169, BC038491, BC071244, and others.

          Matching Taqman Gene Expression Assay:


          Hs00177357_m1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          behavioral fear response
          skeletal muscle contraction
          protein phosphorylation
          apoptotic process
          signal transduction
          germ cell development
          aging
          response to nutrient
          long-term memory
          response to heat
          response to wounding
          response to mechanical stimulus
          response to toxic substance
          response to glucose
          skeletal muscle atrophy
          response to electrical stimulus involved in regulation of muscle adaptation
          positive regulation of smooth muscle cell migration
          cell migration
          TOR signaling
          response to lipopolysaccharide
          response to insulin
          cellular response to hormone stimulus
          response to testosterone
          response to glucagon
          response to tumor necrosis factor
          response to drug
          negative regulation of apoptotic process
          response to leucine
          protein kinase B signaling
          response to ethanol
          positive regulation of translation
          positive regulation of mitotic cell cycle
          positive regulation of translational initiation
          regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of skeletal muscle tissue growth
          positive regulation of smooth muscle cell proliferation
          response to glucocorticoid
          cellular response to growth factor stimulus
          cellular response to organic cyclic compound
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          ribosomal protein S6 kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          kinase activity
          PDZ domain binding
          peptide binding
          identical protein binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0