siRNA Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 3 | 298 | ||
NM_001203245.2 | NP_001190174.1 | 3 | 450 | |||
NM_001204091.1 | NP_001191020.1 | 3 | 302 | |||
GenBank mRNA | AB209165.1 | - | 1 | 253 | ||
AK311797.1 | - | 3 | 245 | |||
BC001032.2 | - | 3 | 223 | 3139393 | ||
BC001955.1 | - | 3 | 277 | 2905318 | ||
BC005012.1 | - | 3 | 277 | 3637658 | ||
BC070235.1 | - | 3 | 221 | 6453788 | ||
BC071946.1 | - | 3 | 233 | 6305105 | ||
BC073799.1 | - | 3 | 310 | 5517455 | ||
BE397113.1 | - | 3 | 248 | |||
BG532491.1 | - | 3 | 451 | |||
BG940847.1 | - | 3 | 298 | |||
BQ072109.1 | - | 1 | 418 | |||
BU956627.1 | - | 3 | 226 | |||
BY996439.1 | - | 3 | 457 | |||
CD385233.1 | - | 3 | 300 | |||
U14972.1 | - | 1 | 220 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 3 | 298 | ||
GenBank mRNA | BU521288.1 | - | 3 | 221 |
Gene Symbol: RPS10P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025353.1 | - | 1 | 2832 |
Gene Symbol: RPS10P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026667.1 | 1 | 354 | |||
GenBank mRNA | AK055590.1 | - | 1 | 1552 | ||
BC030568.2 | - | 1 | 286 |

More Information
Other Species Targeted

Panther Classification:
