Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ighv7-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X66662.1 | - | 2 | 319 |
Gene Symbol: Igh-VS107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M37673.1 | - | 1 | 189 |