Search
Search

ACSM3| Gene Symbol: ACSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005622.3 | NP_005613.2 | 8 | 1305 | ||
| NM_202000.2 | NP_973729.1 | 8 | 1305 | |||
| XM_011545911.1 | XP_011544213.1 | 9 | 1410 | |||
| XM_017023523.1 | XP_016879012.1 | 8 | 1299 | |||
| XM_017023524.1 | XP_016879013.1 | 7 | 1169 | |||
| GenBank mRNA | AK223139.1 | - | 8 | 1274 | ||
| AK290322.1 | - | 8 | 1274 | |||
| AK296122.1 | - | 8 | 1104 | |||
| BC002790.2 | - | 7 | 1164 | 3617826 | ||
| BI826935.1 | - | 2 | 180 | |||
| BU170035.1 | - | 2 | 157 | |||
| D16350.1 | - | 7 | 1145 | |||
| X80062.1 | - | 8 | 1254 | |||