Search
Search

SCD| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005063.4 | NP_005054.3 | 6 | 1398 | ||
| GenBank mRNA | AB032261.1 | - | 6 | 1288 | ||
| AB208982.1 | - | 6 | 1155 | |||
| AF097514.1 | - | 6 | 1143 | |||
| AK222862.1 | - | 6 | 1115 | |||
| AK312312.1 | - | 6 | 1180 | |||
| BC005807.2 | - | 6 | 1085 | |||
| BC062303.1 | - | 6 | 1284 | 6586153 | ||
| CD245516.1 | - | 2 | 49 | |||
| Y13647.1 | - | 6 | 1116 | |||